siRNA Details
| Gene Symbol: Gm19990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045048.1 | 4 | 1053 | |||
| GenBank mRNA | AK053248.1 | - | 4 | 1054 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19990| Gene Symbol: Gm19990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045048.1 | 4 | 1053 | |||
| GenBank mRNA | AK053248.1 | - | 4 | 1054 | ||