siRNA Details
| Gene Symbol: Gm19990 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045048.1 | 1 | 87 | |||
| GenBank mRNA | AK053248.1 | - | 1 | 88 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB664650.1 | - | 1 | 85 | Chr. 12 - 56253245 - 56265813 [+] on Build GRCm38 | |

Gm19990
Genomic Map