siRNA Details
| Gene Symbol: 1700102J08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007112.1 | - | 912 | |||
| AK020344.1 | - | 1 | 114 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721154.1 | - | 1 | 114 | Chr. 7 - 129918790 - 129920269 [-] on Build GRCm38 | |

1700102J08Rik
Genomic Map