siRNA Details
| Gene Symbol: 9330188P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145081.1 | - | 2861 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330188P03Rik| Gene Symbol: 9330188P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145081.1 | - | 2861 | |||