siRNA Details
| Gene Symbol: 9330188P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102319.1 | 1 | 32 | |||
| GenBank mRNA | AK145081.1 | - | 33 | |||
| AK166274.1 | - | 105 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330188P03Rik| Gene Symbol: 9330188P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102319.1 | 1 | 32 | |||
| GenBank mRNA | AK145081.1 | - | 33 | |||
| AK166274.1 | - | 105 | ||||