siRNA Details
| Gene Symbol: 9330188P03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102319.1 | 2 | 376 | |||
| GenBank mRNA | AK034417.1 | - | 1 | 2886 | ||
| AK145081.1 | - | 1913 | ||||
| FJ744746.1 | - | 2 | 298 | |||
| FJ860513.1 | - | 2 | 298 | |||
| FJ860514.1 | - | 2 | 298 | |||

9330188P03Rik
Genomic Map