siRNA Details
| Gene Symbol: Gm5782 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC079904.1 | - | 1 | 191 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF950010.1 | - | 1 | 192 | Chr. 11 - 43414743 - 43416813 [-] on Build GRCm38 | |

Gm5782
Genomic Map