siRNA Details
| Gene Symbol: Gm5106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079527.1 | - | 3 | 1284 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB170853.2 | - | 2 | 601 | Chr. 5 - 8773577 - 8789965 [-] on Build GRCm38 | |

Gm5106
Genomic Map