siRNA Details
| Gene Symbol: 2310041K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009740.1 | - | 1 | 125 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY754692.1 | - | 1 | 136 | Chr. 8 - 71396161 - 71396776 [-] on Build GRCm38 | |

2310041K03Rik
Genomic Map