siRNA Details
| Gene Symbol: 1700012C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005895.1 | - | 2 | 475 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706034.1 | - | 2 | 475 | Chr. 6 - 30460985 - 30463313 [+] on Build GRCm38 | |

1700012C14Rik
Genomic Map