siRNA Details
| Gene Symbol: C030034E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021119.1 | - | 1 | 141 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728247.1 | - | 1 | 138 | Chr. X - 103016128 - 103016618 [+] on Build GRCm38 | |

C030034E14Rik
Genomic Map