siRNA Details
| Gene Symbol: Gm16249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135904.1 | - | 3 | 309 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY719831.1 | - | 3 | 309 | Chr. 13 - 54778086 - 54783091 [-] on Build GRCm38 | |

Gm16249
Genomic Map