siRNA Details
| Gene Symbol: 9530077C14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020648.1 | - | 330 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722269.1 | - | 1 | 330 | Chr. 4 - 139192232 - 139192680 [-] on Build GRCm38 | |

9530077C14Rik
Genomic Map