siRNA Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020640.2 | - | 1 | 335 | ||
| AK142313.1 | - | 1 | 1356 | |||
| AK153781.1 | - | 1 | 1221 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpc4| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020640.2 | - | 1 | 335 | ||
| AK142313.1 | - | 1 | 1356 | |||
| AK153781.1 | - | 1 | 1221 | |||