siRNA Details
Gene Symbol: Gpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020640.2 | - | 1 | 360 | ||
AK142313.1 | - | 1 | 1381 | |||
AK153781.1 | - | 1 | 1246 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020640.2 | - | 1 | 360 | ||
AK142313.1 | - | 1 | 1381 | |||
AK153781.1 | - | 1 | 1246 |