siRNA Details
| Gene Symbol: 1700066B17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040465.1 | 2 | 167 | |||
| GenBank mRNA | BC048628.1 | - | 2 | 167 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU962972.1 | - | 2 | 170 | Chr. 1 - 39842430 - 39847332 [-] on Build GRCm38 | |

1700066B17Rik
Genomic Map