siRNA Details
| Gene Symbol: Gm13483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040361.1 | 3 | 711 | |||
| GenBank mRNA | AK142467.1 | - | 3 | 712 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13483| Gene Symbol: Gm13483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040361.1 | 3 | 711 | |||
| GenBank mRNA | AK142467.1 | - | 3 | 712 | ||