siRNA Details
| Gene Symbol: 4930447K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046184.1 | 3 | 386 | |||
| GenBank mRNA | AK015408.1 | - | 3 | 386 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930447K03Rik| Gene Symbol: 4930447K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046184.1 | 3 | 386 | |||
| GenBank mRNA | AK015408.1 | - | 3 | 386 | ||