siRNA Details
| Gene Symbol: 4933413I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016806.1 | - | 1 | 1221 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV280310.1 | - | 1 | 154 | Chr. 8 - 11726065 - 11726309 [-] on Build GRCm38 | |

4933413I22Rik
Genomic Map