siRNA Details
| Gene Symbol: 1700022A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045509.1 | 1 | 521 | |||
| GenBank mRNA | AK006231.1 | - | 1 | 523 | ||
| AK016801.1 | - | 1 | 549 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700022A22Rik| Gene Symbol: 1700022A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045509.1 | 1 | 521 | |||
| GenBank mRNA | AK006231.1 | - | 1 | 523 | ||
| AK016801.1 | - | 1 | 549 | |||