siRNA Details
| Gene Symbol: 1700022A22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016801.1 | - | 2 | 748 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU937900.1 | - | 1 | 41 | Chr. 15 - 46325758 - 46359893 [+] on Build GRCm38 | |

1700022A22Rik
Genomic Map