siRNA Details
| Gene Symbol: LOC73899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014661.1 | - | 2 | 951 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC73899| Gene Symbol: LOC73899 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014661.1 | - | 2 | 951 | ||