siRNA Details
| Gene Symbol: Gm15270 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014660.1 | - | 3 | 511 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714156.1 | - | 3 | 508 | Chr. 10 - 24592289 - 24595810 [-] on Build GRCm38 | |

Gm15270
Genomic Map