siRNA Details
| Gene Symbol: Wdr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK084217.1 | - | 2 | 1662 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB466570.2 | - | 1 | 565 | Chr. 17 - 14937843 - 14938520 [-] on Build GRCm38 | |

Wdr27
Genomic Map