siRNA Details
| Gene Symbol: 2610030P05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011638.2 | - | 1 | 27 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610030P05Rik| Gene Symbol: 2610030P05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011638.2 | - | 1 | 27 | ||