siRNA Details
Gene Symbol: Tmem208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011635.1 | - | 1 | 226 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY710879.1 | - | 1 | 225 | Chr. 8 - 105326425 - 105327359 [+] on Build GRCm38 |
