siRNA Details
| Gene Symbol: Tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011635.1 | - | 1 | 763 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY710879.1 | - | 1 | 762 | Chr. 8 - 105326425 - 105327359 [+] on Build GRCm38 | |

Tmem208
Genomic Map