siRNA Details
| Gene Symbol: Rab14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043699.1 | - | 1 | 2966 | ||
| AK169946.1 | - | 1 | 2949 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab14| Gene Symbol: Rab14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043699.1 | - | 1 | 2966 | ||
| AK169946.1 | - | 1 | 2949 | |||