siRNA Details
| Gene Symbol: 4930504D19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019668.1 | - | 297 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715236.1 | - | 2 | 297 | Chr. 6 - 84535876 - 84536323 [+] on Build GRCm38 | |

4930504D19Rik
Genomic Map