siRNA Details
| Gene Symbol: 4930505K14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019669.1 | - | 1 | 249 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715248.1 | - | 1 | 241 | Chr. 5 - 148777970 - 148783090 [+] on Build GRCm38 | |

4930505K14Rik
Genomic Map