siRNA Details
| Gene Symbol: Gm19557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045322.1 | 3 | 2742 | |||
| GenBank mRNA | AK158434.1 | - | 3 | 2745 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm19557| Gene Symbol: Gm19557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045322.1 | 3 | 2742 | |||
| GenBank mRNA | AK158434.1 | - | 3 | 2745 | ||