siRNA Details
| Gene Symbol: Agbl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013677.1 | - | 1 | 1637 | ||
| BC028521.1 | - | 4 | 581 | |||
| BC087897.1 | - | 4 | 612 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Agbl3| Gene Symbol: Agbl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013677.1 | - | 1 | 1637 | ||
| BC028521.1 | - | 4 | 581 | |||
| BC087897.1 | - | 4 | 612 | |||