siRNA Details
| Gene Symbol: 4930503F20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019667.1 | - | 2, 3 | 435 | ||
| AK076784.1 | - | 2, 3 | 157 | |||
| BY715229.1 | - | 2, 3 | 435 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930503F20Rik| Gene Symbol: 4930503F20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019667.1 | - | 2, 3 | 435 | ||
| AK076784.1 | - | 2, 3 | 157 | |||
| BY715229.1 | - | 2, 3 | 435 | |||