siRNA Details
Gene Symbol: 4930414N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040294.1 | 2 | 127 | |||
GenBank mRNA | AK015139.1 | - | 2 | 127 | ||
AK076690.1 | - | 2 | 127 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930414N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040294.1 | 2 | 127 | |||
GenBank mRNA | AK015139.1 | - | 2 | 127 | ||
AK076690.1 | - | 2 | 127 |