siRNA Details
| Gene Symbol: 4930414N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040294.1 | 2 | 220 | |||
| GenBank mRNA | AK015139.1 | - | 2 | 220 | ||
| AK076690.1 | - | 2 | 220 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930414N06Rik| Gene Symbol: 4930414N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040294.1 | 2 | 220 | |||
| GenBank mRNA | AK015139.1 | - | 2 | 220 | ||
| AK076690.1 | - | 2 | 220 | |||