siRNA Details
| Gene Symbol: 4930554C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040739.1 | 5 | 1035 | |||
| GenBank mRNA | AK016115.1 | - | 5 | 1037 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930554C24Rik| Gene Symbol: 4930554C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040739.1 | 5 | 1035 | |||
| GenBank mRNA | AK016115.1 | - | 5 | 1037 | ||