siRNA Details
| Gene Symbol: 4930554G24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016119.1 | - | 378 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715721.1 | - | 3 | 378 | Chr. 11 - 14072772 - 14179041 [-] on Build GRCm38 | |

4930554G24Rik
Genomic Map