siRNA Details
| Gene Symbol: D730045B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021342.1 | - | 2 | 805 | ||
| AK080901.1 | - | 845 | ||||
| AK084678.1 | - | 26 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D730045B01Rik| Gene Symbol: D730045B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021342.1 | - | 2 | 805 | ||
| AK080901.1 | - | 845 | ||||
| AK084678.1 | - | 26 | ||||