siRNA Details
| Gene Symbol: D730047P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021347.1 | - | 1 | 261 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730940.1 | - | 1 | 257 | Chr. 9 - 78821880 - 78822371 [+] on Build GRCm38 | |

D730047P14Rik
Genomic Map