siRNA Details
| Gene Symbol: 1700085D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007007.1 | - | 1 | 1058 | ||
| AK054489.1 | - | 1 | 1101 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700085D22Rik| Gene Symbol: 1700085D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007007.1 | - | 1 | 1058 | ||
| AK054489.1 | - | 1 | 1101 | |||