siRNA Details
| Gene Symbol: 1700085D22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007007.1 | - | 1 | 619 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707064.1 | - | 1 | 619 | Chr. 8 - 72548460 - 72549327 [+] on Build GRCm38 | |

1700085D22Rik
Genomic Map