siRNA Details
| Gene Symbol: Rab13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK080805.1 | - | 1 | 20 | ||
| AK083292.1 | - | 1 | 25 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB653712.1 | - | 1 | 22 | Chr. 3 - 90213712 - 90214380 [+] on Build GRCm38 | |

Rab13
Genomic Map