siRNA Details
| Gene Symbol: 4930534H18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005956.1 | - | 1 | 401 | ||
| AK006416.1 | - | 1 | 449 | |||
| AK019718.1 | - | 1 | 458 | |||
| AV040741.2 | - | 1 | 148 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930534H18Rik| Gene Symbol: 4930534H18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005956.1 | - | 1 | 401 | ||
| AK006416.1 | - | 1 | 449 | |||
| AK019718.1 | - | 1 | 458 | |||
| AV040741.2 | - | 1 | 148 | |||