siRNA Details
| Gene Symbol: 4930534H18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005956.1 | - | 1 | 254 | ||
| AK006416.1 | - | 1 | 302 | |||
| AK019718.1 | - | 1 | 311 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930534H18Rik| Gene Symbol: 4930534H18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005956.1 | - | 1 | 254 | ||
| AK006416.1 | - | 1 | 302 | |||
| AK019718.1 | - | 1 | 311 | |||