siRNA Details
| Gene Symbol: Kcnd3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040759.1 | 2 | 444 | |||
| GenBank mRNA | AK018929.1 | - | 98 | |||
| AK131816.1 | - | 2 | 444 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707553.1 | - | 2 | 439 | Chr. 3 - 105451270 - 105452950 [-] on Build GRCm38 | |

Kcnd3os
Genomic Map