siRNA Details
| Gene Symbol: 2810414N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013093.1 | - | 1 | 354 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CX203092.1 | - | 1 | 595 | Chr. 1 - 151749453 - 151750231 [-] on Build GRCm38 | |

2810414N06Rik
Genomic Map