siRNA Details
| Gene Symbol: Gm34240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085621.1 | - | 3 | 551 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm34240| Gene Symbol: Gm34240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085621.1 | - | 3 | 551 | ||