siRNA Details
| Gene Symbol: Gm32960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK085625.1 | - | 1 | 32 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB661619.1 | - | 1 | 32 | Chr. 19 - 38348702 - 38349608 [+] on Build GRCm38 | |

Gm32960
Genomic Map