siRNA Details
| Gene Symbol: 5330439K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019939.1 | - | 2 | 1247 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

5330439K02Rik| Gene Symbol: 5330439K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019939.1 | - | 2 | 1247 | ||