siRNA Details
| Gene Symbol: 5730419F03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017575.1 | - | 350 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY717356.1 | - | 2 | 347 | Chr. 1 - 78596263 - 78609506 [-] on Build GRCm38 | |

5730419F03Rik
Genomic Map